ContactUsLinkComponent
VWR* Lens Cleaning Tissues
VWR® Lens Cleaning Tissues
# de catalogue 52846-001
Fournisseur:  VWR International
Product available on GSA Advantage®
VWR* Lens Cleaning Tissues
VWR® Lens Cleaning Tissues
# de catalogue 52846-001
Fournisseur:  VWR International
Product available on GSA Advantage®
Spécifications
  • Dimensions:
    10.1 x 15.2 cm (4 x 6")
  • Cat. no.:
    52846-001
  • Packaging:
    50 Sheets/Book, 12 Books/Pack
Spécifications
A propos de cet article
Dust-free lens tissues are ideal for use with liquid cleaners.
  • High wet strength
Clean safety glasses and goggles, microscope lenses, and more with no scratching.
Customers Who Bought This Also Bought