ContactUsLinkComponent
VWR* Lens Cleaning Tissues
VWR® Lens Cleaning Tissues
# de catalogue 52845-009
Fournisseur:  VWR International
VWR® Lens Cleaning Tissues
# de catalogue 52845-009
Fournisseur:  VWR International
Spécifications
  • Dimensions:
    15.2 x 20.3 cm (6 x 8")
  • Cat. no.:
    52845-009
  • Packaging:
    50 Sheets/Book, 12 Books/Pack
Spécifications
A propos de cet article
Dust-free lens tissues are ideal for use with liquid cleaners.
  • High wet strength
Clean safety glasses and goggles, microscope lenses, and more with no scratching.
Customers Who Bought This Also Bought