To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Range:0–30 mg/dL
- Resolution:0.1 mg/dL
- Sample size:30 µL
- Description:Total Bilirubin Test Kit
- Electrical:100–130V, 60Hz, 20W
- Shipping weight:5.5 kg (12 lbs.)
- Results displayed, total:Instant
- Results displayed, direct:2 min.
- Repeatability:±0.2 mg/dL (1 S.D.)
- Cat. no.:52197-566
- Supplier No.:B2A101
Spécifications
A propos de cet article
Analyzer provides a rapid and accurate determination of total and direct bilirubin in neonates from a single 30µL serum sample
Using spectrophotometric analysis, total bilirubin level is determined immediately and direct bilirubin level is determined in about two minutes. Analyzer is easy to use, with simple set up, calibration, and operation.
Dimensions: 330×280×180 mm (13×11×7")