ContactUsLinkComponent
Kjeldahl Granules, Selenized
Kjeldahl Granules, Selenized, Hengar
# de catalogue 51137-046
Fournisseur:  Hengar
Small Business Enterprise
Numéro CAS:  
Kjeldahl Granules, Selenized
Kjeldahl Granules, Selenized, Hengar
# de catalogue 51137-046
Fournisseur:  Hengar
Small Business Enterprise
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    250 g
  • Cat. no.:
    51137-046
  • Supplier No.:
    901200
Spécifications
A propos de cet article
Granules greatly reduce time required for the Kjeldahl digestion, and ensure smooth boiling, both during digestion and subsequent distillation
Selenium is applied as a coating on the granules, as a convenient means to add to the digestion. Only two or three granules for each digestion flask is necessary.
Customers Who Bought This Also Bought