Spécifications
- Size:250 g
- Cat. no.:51137-046
- Supplier No.:901200
Spécifications
A propos de cet article
Granules greatly reduce time required for the Kjeldahl digestion, and ensure smooth boiling, both during digestion and subsequent distillation
Selenium is applied as a coating on the granules, as a convenient means to add to the digestion. Only two or three granules for each digestion flask is necessary.