Kimble® Disposable Pasteur Pipettes, Soda Lime, DWK Life Sciences
# de catalogue 50880-040
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Length:14.6 cm
- Description:Soda Lime Monstr-Pette Pasteur Pipet
- Cat. no.:50880-040
- Supplier No.:63DP1005
Spécifications
A propos de cet article
These pipettes feature straight-cut tips and smooth, fire-polished tops to facilitate the attachment of bulbs.
- Made of soda-lime glass
- Available with cotton-plugged or unplugged
They are excellent for use in blood banks, hematology, chemistry, bacteriology and other general lab applications.
Customers Who Bought This Also Bought