Order Entry
ContactUsLinkComponent
Mortar and Pestle Set, Glass
Mortar and Pestle Sets, Glass, Variety Glass
# de catalogue 50415-085
Fournisseur:  Variety Glass
Mortar and Pestle Set, Glass
Mortar and Pestle Sets, Glass, Variety Glass
# de catalogue 50415-085
Fournisseur:  Variety Glass

Spécifications

  • Volume:
    455 ml
  • Height:
    120 mm
  • Inner Diameter:
    135 mm
  • Supplier No.:
    826-5

Spécifications

A propos de cet article

Mortar and pestle sets are made of clear, heavy, flint glass

Mortar has a heavy molded base and lip for pouring. Sold as a set.

Customers Who Bought This Also Bought