Spécifications
- Description:Microscope Slide Holders
- Cat. no.:48457-107
Spécifications
À propos de cet article
Cabinets are made of steel and are ideal for organizing slides.
- Steel fabrication
- Compatible with all slides
The single drawer cabinet stores 100 micro slide holders (up to 400 slides)
Index card micro slide holders offer instant reference and fast fingertip retrieval. They sort, classify, or arrange slides in any order. Cards can be added, removed, or changed for fast updating. Durable holder is 3x5" cardboard fitted with a protective plastic frame, which provides secure protection of specimens. Each slide holder card accepts four 25x75mm (1x3") slides of any thickness. Visible four-position tabs provide “in-sight” indexing. Ruled data lines are keyed to numbered slots and allow complete slide identification and comments.
The double drawer cabinet stores 200 slide holders (up to 800 slides).