VWR® Microscope Slides and Plastic Coverslips
Catalog # 48376-049
Supplier: VWR International
Specifications
- Description:Plastic cover slips
- L×W:22×22 mm
- Thickness:0.25 mm
- Cat. no.:48376-049
- Packaging:10 inner packs of 100
Specifications
About this item
Efficient, disposable slides are manufactured from optically clear vinyl.
- Made of clear vinyl plastic
- Refractive index as glass
- Unbreakable and safer than glass
- Used in biological and medical labs, as well as educational applications
Slides will not break, scratch, or cut fingers. Ideal for wet mounts. Micro coverslips are unbreakable and non-curling.
Customers Who Bought This Also Bought