BASE ALUMINUM BOTTLE CYLINDER 2IN CYL
# de catalogue 47752-772
Fournisseur: Airgas
Customers Who Bought This Also Bought
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
79,43 $ to 662,05 $
WHEATON® MicroLiter Screw-Thread Vials, Component Kits, Assembled, 9 mm, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
127,72 $
RSA™ and RSA-Pro™ Autosampler Vials, Caps and Convenient Kits
Fournisseur: MicroSolv Technology Corp
433,35 $
SureSTART™ Septa for 20 mm Stoppers, Level 2 High-throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
1 294,78 $ to 1 294,78 $
SureSTART™ GOLD-Grade Glass Insert for 2 ml Vials, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
810,00 $ to 840,15 $
WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
205,86 $
WHEATON® MicroLiter 9 mm Screw-Thread Vials, Component Kits, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
76,34 $
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
149,40 $
SureSTART™ Glass Inserts for 2 ml Vials, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
120,60 $ to 125,07 $
WHEATON® MicroLiter µLPlate® Component Kits, Inserts and Caps, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
184,38 $