REGULATOR CGA540 ANAL CYL 1STAGE 0 25PSI
# de catalogue 47752-344
Fournisseur: Airgas
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
195,27 $
WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
343,64 $
Screw Top Vials and Caps, Easy Purchase Packs 9 mm, MicroSolv
Fournisseur: MicroSolv Technology Corp
129,58 $
WHEATON® MicroLiter 9 mm Screw-Thread Vials, Component Kits, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
167,84 $
MicroSolv™ Pharma Grade Vials, Caps and Convenience Kits
Fournisseur: MicroSolv Technology Corp
155,33 $
SureSTART™ Glass Screw Top 0.3 ml Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
182,38 $ to 196,03 $
SureSTART™ Polypropylene Screw Top 2 ml Microvials for <2 ml Samples, Level 1 Everyday Analysis, Thermo Scientific
Fournisseur: Thermo Scientific
390,18 $ to 390,18 $