REGULATOR ANAL 1STAGE NDL CGA 580 50PSI
# de catalogue 47752-058
Fournisseur: Airgas
Customers Who Bought This Also Bought
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$78.90 to $874.91
Water ACS, Reagent Grade ASTM Type I, ASTM Type II Packaged in plastic containers
Fournisseur: Ricca Chemical
$200.03
RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
Fournisseur: Thermo Scientific
$607.92 to $2,080.63
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
$635.20 to $2,541.50
1-Octanesulfonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
$814.96
Ion Chromatography (IC) Anion and Cation Check Standards, VWR Chemicals BDH®
Fournisseur: VWR International
$306.90