REGULATOR ANAL 1STAGE CGA 350 0 25 PSI
# de catalogue 47752-050
Fournisseur: Airgas
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
120,43 $ to 1 204,30 $
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
46,59 $ to 465,92 $
SureSTART™ Glass Inserts for 2 ml Vials, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
120,60 $ to 125,07 $
AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
Fournisseur: MicroSolv Technology Corp
54,30 $
SureSTART™ Screw Caps, 9 mm, Level 3 High Performance Applications
Fournisseur: Thermo Scientific
79,21 $ to 82,14 $
WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
210,42 $