REGULATOR 1STAGE BRASS ANAL LINE 0 25PSI
# de catalogue 47752-044
Fournisseur: Airgas
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$118.61 to $1,128.11
MicroSolv™ Pharma Grade Vials, Caps and Convenience Kits
Fournisseur: MicroSolv Technology Corp
$52.50
SureSTART™ GOLD-Grade Glass Insert for 2 ml Vials, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$810.00 to $840.15
RSA™ and RSA-Pro™ Autosampler Vials, Caps and Convenient Kits
Fournisseur: MicroSolv Technology Corp
$438.81
WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$205.86
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$176.80
SureSTART™ Glass Inserts for 2 ml Vials, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$120.60 to $125.07