Order Entry
ContactUsLinkComponent
Box, Storage
SP Bel-Art ProCulture Cryogenic Vial Storage Boxes, Bel-Art Products, a part of SP
Catalog # 47751-740
Box, Storage
SP Bel-Art ProCulture Cryogenic Vial Storage Boxes, Bel-Art Products, a part of SP
Catalog # 47751-740

Specifications

  • No. of Holes:
    81
  • Description:
    9 x 9 Vial Storage Box
  • Dimensions:
    13.3L×13.3W×5.1H cm (5¼×5¼×2”)
  • Cat. no.:
    47751-740
  • For Vials:
    1.2 - 2.0 ml
  • Supplier No.:
    188490001

Specifications

About this item

Durable polycarbonate boxes are ideal for the compact storage of cell stocks in 1.2, 2.0, or 5.0 mL cryogenic vials.

  • Can be used in temperatures from –196 ºC to +121 ºC
  • Clear lids can be written on with permanent markers and have a printed grid for easy inventory
  • Keyed corner aligns the lid with identifying numbers on the rack
  • Useful as mailers for vials

Box 47751-740 can be used with Cryo Tower Storage Systems (74950-132 and 74950-134) to save valuable bench space.

Customers Who Bought This Also Bought