Order Entry
ContactUsLinkComponent
Polygon Spinbar* Magnetic Stirring Bar
VWR® Polygon Spinbar Magnetic Stirring Bars without Pivot Ring
# de catalogue 47751-482
Polygon Spinbar* Magnetic Stirring Bar
VWR® Polygon Spinbar Magnetic Stirring Bars without Pivot Ring
# de catalogue 47751-482

Spécifications

  • Ø:
    6 mm (1/4")
  • Length:
    15 mm (5/8")
  • Cat. no.:
    47751-482
  • Shape:
    Polygon
  • Supplier No.:
    371205015

Spécifications

A propos de cet article

These stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.

  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert
  • Autoclave safe

Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.

Customers Who Bought This Also Bought