Order Entry
ContactUsLinkComponent
FILTER PAPER
Wet-Strengthened Qualitative Filter Paper, Grade 909, Ahlstrom
# de catalogue 47749-728
Fournisseur:  Ahlstrom
FILTER PAPER
Wet-Strengthened Qualitative Filter Paper, Grade 909, Ahlstrom
# de catalogue 47749-728
Fournisseur:  Ahlstrom

Spécifications

  • Ø:
    18.5 cm
  • Flow rate:
    Medium
  • Retention:
    3.0 µm
  • Load capacity:
    Medium
  • Cat. no.:
    47749-728
  • Supplier No.:
    9090-1850

Spécifications

A propos de cet article

Ahlstrom Grade 909 has a rough surface, a medium flow rate, and is a tighter version of Grade 962.

Customers Who Bought This Also Bought