ContactUsLinkComponent
Coupling, 2.54cm (1") Beaded Pipe
Accessories for Jacketed Cylindrical Reaction System, Chemglass
Catalog # 47733-950
Supplier:  Chemglass
Small Business Enterprise
Coupling, 2.54cm (1") Beaded Pipe
Accessories for Jacketed Cylindrical Reaction System, Chemglass
Catalog # 47733-950
Supplier:  Chemglass
Small Business Enterprise
Specifications
  • Description:
    1" Beaded Pipe Coupling (2)
  • Cat. no.:
    47733-950
  • Supplier No.:
    CG-1968-66