Order Entry
ContactUsLinkComponent
Dessicant
Desiccant Packs, Desco
# de catalogue 47731-930
Fournisseur:  Desco
Desiccant Packs, Desco
# de catalogue 47731-930
Fournisseur:  Desco

Spécifications

  • Dimensions:
    3.8 x 7.6 cm (1.5 x 3")
  • Description:
    Desiccant Packs, Pail of 550
  • Cat. no.:
    47731-930
  • Supplier No.:
    13850

Spécifications

A propos de cet article

Drying agent used to lower the moisture content of air inside a closed space, such as an MBB bag.

Certifications: Meets MIL-D-3464, Type II
Customers Who Bought This Also Bought