Order Entry
ContactUsLinkComponent
Thermal Paper
VWR® Thermal Printer Paper
# de catalogue 47729-740
Fournisseur:  VWR International
VWR® Thermal Printer Paper
# de catalogue 47729-740
Fournisseur:  VWR International

Spécifications

  • Description:
    Thermal printer paper
  • Cat. no.:
    47729-740
  • Packaging:
    1 Roll
  • Length:
    18 mm
  • Width:
    58 mm

Spécifications

A propos de cet article

Heat sensitive paper for use with VWR® sterilizer with printer model AS12P.

Customers Who Bought This Also Bought