ContactUsLinkComponent
Utility Tray
SP Bel-Art Utility Trays, Polypropylene, Bel-Art Products, a part of SP
# de catalogue 47728-018
Utility Tray
SP Bel-Art Utility Trays, Polypropylene, Bel-Art Products, a part of SP
# de catalogue 47728-018
Spécifications
  • Cat. no.:
    47728-018
  • L×W×H:
    40.6×66×7.6 cm (16×26×3")
  • Supplier No.:
    800160237A
Spécifications
A propos de cet article
Made from polypropylene, these straight-sided trays offer resistance to temperatures up to 135 °C (274 °F).
  • Polypropylene construction
  • Good temperature resistance
  • Straight-sided design
  • Autoclavable
Customers Who Bought This Also Bought