ContactUsLinkComponent
Disposable Culture Tubes
KIMAX® Culture Tubes, Disposable, Soda-Lime Glass, Kimble Chase
# de catalogue 46610-182
Disposable Culture Tubes
KIMAX® Culture Tubes, Disposable, Soda-Lime Glass, Kimble Chase
# de catalogue 46610-182
Spécifications
  • Capacity:
    11 ml
  • Ø:
    16 mm
  • Description:
    Plain Disposable Soda-Lime Glass Tubes
  • Height:
    75 mm
  • Supplier No.:
    60MM190
Spécifications
A propos de cet article
Plain disposable soda-lime glass tubes manufactured from 90 expansion soda-lime glass conforming to USP Type III requirements.
  • All tubes have sturdy, uniform bottoms and consistent lengths
  • No marking spot
Customers Who Bought This Also Bought