PIG® Dri Loose Absorbent, New Pig
# de catalogue 46000-498
Fournisseur: New Pig
Spécifications
- Cat. no.:46000-498
- Packaging:50 x 40 lb. Bags
- Supplier No.:PLP213-50
Spécifications
A propos de cet article
Unlike clay, which is simply coated with liquid, fast-wicking cellulose-based kaolin granules begin to soak up liquid the moment they touch the spill.
- Absorbs and retains oils, coolants, solvents and water
- Can be incinerated after use to reduce waste or for fuels blending
100% recycled content is a byproduct of the paper-making process, Excellent when green products are desired or required. Silica-free composition does not pose the health hazards associated with breathing in clay dust. Light-weight absorbent is easier than clay to carry and use without injury.
Customers Who Bought This Also Bought