Spécifications
- Clothing Size:XL
- Garment application:Laboratory
- Color:Blue
- Weight:50.45 g/m²
- Cat. no.:414004-403
Spécifications
A propos de cet article
Frocks are manufactured from a multilayered spunbonded-meltblown-spunbonded (SMS) fabric that offers high tensile strength while remaining soft, comfortable, and breathable.
- Multilayer SMS material protection
- Light fluid and particulate barrier
- Excellent breathability
- No pockets
- Serged seams
These static-dissipative garments act as a fluid and particulate barrier without sacrificing comfort or restricting movement
Garments feature snap collar and snap front, raglan sleeves, and elastic cuffs. No pockets. Serged seams.
Frocks are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
They provide light chemical splash resistance. The material is low-linting and resistant to tears and punctures. Ideally suited for use in high-class protection areas of medical, industrial, laboratory, electronic, and pharmaceutical environments.