ContactUsLinkComponent
VWR LABCT MAXP 4XL WHITE CS25
VWR® Maximum Lab Coats
# de catalogue 414004-374
Fournisseur:  VWR International
VWR LABCT MAXP 4XL WHITE CS25
VWR® Maximum Lab Coats
# de catalogue 414004-374
Fournisseur:  VWR International
Spécifications
  • Clothing Size:
    4XL
  • Garment application:
    Laboratory
  • Color:
    White
  • Weight:
    63 g/m²
  • Cat. no.:
    414004-374
Spécifications
A propos de cet article
These lab coats are the cleanest and most durable available within the VWR® Protection line.
  • Superior fluid and particulate barrier
  • Lowest level of particle shedding
  • Light chemical-splash resistance
  • Breathable to ensure user comfort
  • Soft, cloth-like fabric with maximum tear strength
Manufactured from a specially formulated breathable microporous fabric, these garments are distinguished by their exceptional comfort benefits and superior barrier protection capabilities
Garments feature tapered collar, snap front, inset sleeves, elastic cuffs, and three pockets. Serged seams.
The lab coats provide barrier protection against fluids, biologicals, particulates, and light chemical splash. A high vapor transmission rate keeps the wearer comfortable, even throughout extended use. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental cleanup applications.
Certifications: Lab coats are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
Customers Who Bought This Also Bought