VWR® Disposable Aspirating Pipettes, Polystyrene, Sterile
# de catalogue 414004-268
Fournisseur: VWR International
Spécifications
- Capacity:25 ml
Spécifications
A propos de cet article
VWR® Disposable Aspirating Pipettes are manufactured from polystyrene and supplied sterile.
- Non-plugged, nongraduated, single-use pipettes
- Thermoformed paper/plastic packaging protects against contamination
These non-plugged, nongraduated, single-use pipettes can be used for a wide variety of vacuum-aspirating procedures, and are a safer alternative to glass pasteur pipettes. Pipettes are supplied in a thermoformed paper/plastic packaging wrap to protect against contamination. Nonpyrogenic.
Packaging: Pipettes are supplied individually wrapped in four bags of 50.
Customers Who Bought This Also Bought