Accessories for UVP Blak-Ray® and Sterilizing UV Bench/Display Lamps, Analytik Jena
# de catalogue 36585-280
Fournisseur: Analytik Jena US
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Replacement Tube
- For Model:XX-15BLB
- Wavelength:365 nm
- Supplier No.:34-0017-01