Accessories for UVP PL Series Compact and Handheld Ultraviolet Lamps, Analytik Jena
# de catalogue 36584-107
Fournisseur: Analytik Jena US
Product available on GSA Advantage®
Spécifications
- Description:Lamp Stand, Model J124, for 4W Compact Lamps, 165H×190W×122D mm
- Cat. no.:36584-107
- For Use With:UVG-11, UVL-21, UVGL-25
- Supplier No.:98-0020-03
Spécifications
A propos de cet article
Accessories include replacement tubes, filter assemblies, lamp stands, batteries, and chargers.
Customers Who Bought This Also Bought