Order Entry
ContactUsLinkComponent
Large, Heavy-Duty Polyethylene Scoop
VWR® Large Cylindrical Scoops, PE
Catalog # 36494-110
Supplier:  VWR International
Large, Heavy-Duty Polyethylene Scoop
VWR® Large Cylindrical Scoops, PE
Catalog # 36494-110
Supplier:  VWR International

Specifications

  • Capacity:
    4L (1 gal.)
  • Cat. no.:
    36494-110
  • Handle Length:
    12.7 cm
  • Bowl Dimensions (W×L×H):
    19.1×24.8 cm

Specifications

About this item

Large, heavy-duty, polyethylene plastic scoops are strong, durable, and excellent for scooping dry powders, granules, or crystals.

  • Closed back, non-spill shape
  • Ergonomically angled handle
Customers Who Bought This Also Bought