Barrier Tapes, National Marker
# de catalogue 36434-305
Fournisseur: ACCUFORM MANUFACTURING, INC
Spécifications
- W×L:5.1 cm x 45.7 m (2" x 50 yds.)
- Color:Red/Black Stripe
- Description:Striped barrier tapes
- Cat. no.:36434-305
- Supplier No.:BT7RB
Spécifications
A propos de cet article
Use to mark off hazardous areas. Utilize as you would a rope.
- Durable, lightweight polyethylene
- Red-orange barrier tape offers 4X the visibility of other colors
- Reusable, non-adhesive, chemical resistant, weatherproof, fade resistant
Printed barricade tapes are made of durable, lightweight polyethylene
Fluorescent red-orange barrier tape offers four times the visibility of other colors. Reusable, nonadhesive tape is resistant to most chemicals. Two-color, striped barrier tapes are made of woven polypropylene for extra durability. Chemical-resistant, weatherproof, and fade-resistant tape can be reused many times.
Printed legends warn of potential hazards and dangerous working areas.
Customers Who Bought This Also Bought