VWR® Write-On Metal Tags
# de catalogue 36419-213
Fournisseur: VWR International
Spécifications
- Description:Write-on metal tags
- Material:Metal
- Cat. no.:36419-213
- Dimensions:34.9×79.5 mm (1.38×3.13")
Spécifications
A propos de cet article
Permanently engrave your message on this metal tag with an ordinary ballpoint pen.
- Extremely durable
- Suited to extreme hot or cold temperatures
Permanently engrave your message on this metal tag with an ordinary ballpoint pen.
The aluminum tag features a 'soft center' and will take messages on both sides.
Delivery: Package of 48 tags with wire fasteners.
Customers Who Bought This Also Bought