Order Entry
Canada
ContactUsLinkComponent
Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor, Thermo Scientific
Catalogue # 34182-070
Numéro CAS:  
undefined
Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor, Thermo Scientific
Catalogue # 34182-070
Supplier Number:  700702
Numéro CAS:  

Spécifications

  • Description:
    Buffer A, pH 6.97
  • Cat. no.:
    34182-070
  • Supplier No.:
    700702

Spécifications

À propos de cet article

Sample well water, rain water, distilled/deionized water, boiler feed water, process water, and more with the Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor

  • Increases measurement accuracy
  • Reduces electrode drift
  • Eliminates contamination of sample
  • For best results, a high quality pH electrode is recommended

It offers fast, accurate pH results in high purity and low ionic strength waters.

Ordering information: Kit contains special low ionic strength buffers for calibration and pHISA™ adjustor additive, to increase sample ionic strength without changing pH.
Frequently Bought Together