To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:pH 10.01 Buffer
- Size:10 x 15 mL
- Cat. no.:34175-514
- Supplier No.:911010
Spécifications
A propos de cet article
Ensure fresh, ready to use standards for pH calibration with Orion™ pH Buffer Individual Use Pouches
- Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, and pH 10.01 buffer is blue
- Packaged with buffer/temperature tables for accurate calibrations and NIST traceability
- Manufactured under ISO 9000 quality standards
- Pouches are 15mL each and are available in 10 packs or 25 packs
Each pouch is sealed against light, air, or other contamination, and is disposable. There is no wasted buffer or secondary reservoir. Each pouch contains lot code, expiration date, and buffer temperature table.
Buffers are color coded for ease of use. All Orion™ pH buffers are traceable to NIST standard reference materials.