Order Entry
ContactUsLinkComponent
PerpHecT* pH 4 Buffer
Orion™ pH Buffer Individual Use Pouches, Thermo Scientific
Catalog # 34175-510
CAS Number:  
PerpHecT* pH 4 Buffer
Orion™ pH Buffer Individual Use Pouches, Thermo Scientific
Catalog # 34175-510
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    pH 4.01 Buffer
  • Size:
    10 x 15 mL
  • Cat. no.:
    34175-510
  • Supplier No.:
    910410-WA

Specifications

About this item

Ensure fresh, ready to use standards for pH calibration with Orion™ pH Buffer Individual Use Pouches

  • Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, and pH 10.01 buffer is blue
  • Packaged with buffer/temperature tables for accurate calibrations and NIST traceability
  • Manufactured under ISO 9000 quality standards
  • Pouches are 15mL each and are available in 10 packs or 25 packs

Each pouch is sealed against light, air, or other contamination, and is disposable. There is no wasted buffer or secondary reservoir. Each pouch contains lot code, expiration date, and buffer temperature table.

Buffers are color coded for ease of use. All Orion™ pH buffers are traceable to NIST standard reference materials.

Customers Who Bought This Also Bought