Spécifications
- Description:Optimum Result™ B Fill solution for chloride, cyanide, lead and silver/sulfide ISE
- Size:5×60 ml
- Cat. no.:34117-965
- For:9682BNWP combination or 9482BN and 900200 half-cell electrodes
- Supplier No.:900062
Spécifications
A propos de cet article
Enhance the performance and extend the life of ion selective electrodes with Orion™ ISE Filling Solutions.
- Filling/maintenance solutions are for use with Orion™ Ion Selective Electrodes (ISE)
- Optimum Result™ ISE filling solutions are specially designed for the Ionplus™ series ion selective electrodes
These solutions are specially formulated to work with the recommended ion selective electrodes and prevent electrode interferences from filling solution.