ContactUsLinkComponent
Filling Solution
Orion™ ISE Filling Solutions, Thermo Scientific
# de catalogue 34113-948
Numéro CAS:  
Filling Solution
Orion™ ISE Filling Solutions, Thermo Scientific
# de catalogue 34113-948
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Fill solution for sodium ISE
  • Size:
    5×60 ml
  • Cat. no.:
    34113-948
  • For:
    8611BNWP combination or 8411BN and 800300 half-cell electrodes
  • Supplier No.:
    900010
Spécifications
A propos de cet article
Enhance the performance and extend the life of ion selective electrodes with Orion™ ISE Filling Solutions.
  • Filling/maintenance solutions are for use with Orion™ Ion Selective Electrodes (ISE)
  • Optimum Result™ ISE filling solutions are specially designed for the Ionplus™ series ion selective electrodes
These solutions are specially formulated to work with the recommended ion selective electrodes and prevent electrode interferences from filling solution.
Customers Who Bought This Also Bought