Spécifications
- Description:Replacement module for potassium half-cell electrode
- Cat. no.:34113-416
- Supplier No.:931901
Spécifications
A propos de cet article
Measure potassium in aqueous solutions simply, accurately, and economically with Orion™ Potassium Electrodes.
- Free-flowing Sure-Flow junction assures stable, drift-free potentials
A separate reference electrode is unnecessary, making this electrode convenient to use with small sample volumes.