ContactUsLinkComponent
Plastic Membrane Electrode Modules for Potassium
Orion™ Potassium Electrodes, Thermo Scientific
# de catalogue 34113-416
Orion™ Potassium Electrodes, Thermo Scientific
# de catalogue 34113-416
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Replacement module for potassium half-cell electrode
  • Cat. no.:
    34113-416
  • Supplier No.:
    931901
Spécifications
A propos de cet article
Measure potassium in aqueous solutions simply, accurately, and economically with Orion™ Potassium Electrodes.
  • Free-flowing Sure-Flow junction assures stable, drift-free potentials
A separate reference electrode is unnecessary, making this electrode convenient to use with small sample volumes.
Customers Who Bought This Also Bought