ContactUsLinkComponent
Orion™ pH Electrode Filling Solution, Thermo Scientific
Catalogue # 34108-020
Numéro CAS:  
undefined
Orion™ pH Electrode Filling Solution, Thermo Scientific
Catalogue # 34108-020
Supplier Number:  900011
Numéro CAS:  
Spécifications
  • Description:
    Fill Solution for Standard, Speciality, PerpHecT Ag/AgCl, Single Junction Refillable Green and Micro pH Electrodes, ORP Electrodes
  • Size:
    60 mL
  • Cat. no.:
    34108-020
  • Supplier No.:
    900011
Spécifications
À propos de cet article
Use Orion™ pH Electrode Filling Solution with pH electrodes and reference electrodes.
Only use ROSS filling solution with ROSS™ and ROSS Ultra™ pH electrodes to prevent damage. When the single junction reference electrode or double junction reference electrode are used with Orion™ ion selective electrodes (ISE), refer to the recommended ISE filling solution listed in the ISE user guide.
Frequently Bought Together