Visorgogs® Safety Goggles, Jones FR
# de catalogue 33012-006
Fournisseur: Jones & Co.
Spécifications
- Description:Indirect vent goggles
- Supplier No.:34862-BLUE
Spécifications
A propos de cet article
These full vision safety goggles are specially designed to protect eyes from flying particles or splashing liquids.
- Indirect ventilation screen keeps dirt and dust out and keeps lenses fog-free
- Lenses are designed strutless with a down-angle vision for clear visibility
- Goggles can be worn over eyeglasses
- Available in four visor colors with clear lenses
- Replacement lens fits both indirect and direct vent goggles
Unique design incorporates comfortable, impact-resistant goggles with a vented, opaque visor to shade eyes and reduce eye strain and fatigue.
Certifications: Meet ANSI Z87.1 standard.
Customers Who Bought This Also Bought