UVP Blak-Ray® Safety Goggles, Analytik Jena
Catalog # 33002-078
Supplier: Analytik Jena US
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:UVC-503 goggles, UV blocking, 76H×152W mm
- Cat. no.:33002-078
- Supplier No.:98-0002-02
Specifications
About this item
Goggles provide maximum protection from exposure to ultraviolet wavelengths and reduce blue haze interference.
They feature replaceable lenses. Lightweight design and flexible rubber trim provide a comfortable fit, even over prescription glasses. Air vents allow air flow while blocking ultraviolet light.
Certifications: Meet NIOSH, OSHA, and ANSI specifications. CE approved.
Frequently Bought Together