PYREX® Capillary Gas Delivery Tube, Corning
Catalog # 32638-915
Supplier: Corning
Specifications
- Description:PYREX® Capillary Gas Delivery Tube, 14/10
- Cat. no.:32638-915
- Supplier No.:9070-14
Specifications
About this item
This capillary gas delivery tube is a replacement part for the 6949M series of PYREX® Microchemistry Kits.
Customers Who Bought This Also Bought