Order Entry
ContactUsLinkComponent
Costar* Brand 96-Well Plates, Polypropylene, Sterile, Without Lids
Corning® 96-Well Plates, Polypropylene, Without Lids
Catalog # 29444-400
Supplier:  Corning
Product available on GSA Advantage®
Corning® 96-Well Plates, Polypropylene, Without Lids
Catalog # 29444-400
Supplier:  Corning
Product available on GSA Advantage®

Specifications

  • Sterility:
    Sterile
  • Well Volume:
    320 µl
  • Color:
    Clear
  • Description:
    Storage plate, 96-well, natural, V-shaped well
  • Cat. no.:
    29444-400
  • Well Shape:
    V-Bottom
  • Supplier No.:
    3357

Specifications

About this item

Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.

  • Flat, round, or V-shaped well bottom
  • Features uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • RNase-/DNase-free
  • Available sterile or non sterile
Ordering information: Plates are packaged 25 per pack, 4 packs per case. Nonsterile 450 µl plate is packaged 10 plates per bag and 5 bags per case.
Customers Who Bought This Also Bought