Cell Culture Storage Bottles, Polystyrene, Corning
# de catalogue 29442-870
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Capacity:125 ml (4.2 oz.)
- Cat. no.:29442-870
- Supplier No.:8388
Spécifications
A propos de cet article
Disposable polystyrene bottles for storage of media, buffers and other aqueous solutions.
- Low profile, easy grip style has sides that facilitate handling
- Plug seal caps (45 mm) provide an airtight seal and help minimise the risk of contamination
- Bottles can be used with Corning® vacuum filter systems
- Sterile, certified non-pyrogenic
Customers Who Bought This Also Bought