Universal Fit Pipet Tips, Corning®
1 - 200 µl, +, Naturel, Rack Packed
Supplier: Corning
Fournisseur #: 4864
Universal Fit Pipet Tips, Corning®
Catalog # 29442-684
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
Corning® universal fit tips are designed to provide a reliable fit with all major brand pipettors.
- Beveled orifice helps ensure accurate fluid delivery
- All tips are certified RNase-/DNase-free and non pyrogenic
- Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design
- Bulk packed tips are non sterile and very economical
1 to 200 µl universal fit tips are graduated at the 10 µl, 50 µl, and 100 µl volumes.
Spécifications
- Volume:1 - 200 µl
- Tip Color:Naturel
- Packed:Rack Packed
- Sterile:+
- Cat. No.:29442-684
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Environmentally Preferable:Low Manufacturing Impact