Corning® Gel-Loading Pipette Tips, Corning
Flat, O.D. 0.4 mm, 0.1 - 200 µl
: Corning
: 4854
These gel-loading pipette tips feature a capillary end that allows easy access into vertical and horizontal electrophoresis gels.
- Disposable
- Certified RNase-/DNase-free and non pyrogenic
: Available in flat and round tip configurations. Flat tips are designed for use in 0.2 and 0.4 mm sequencing gels. Round tips are designed for use in SDS-PAGE 0.5 mm gels.
- Volume:0.1 - 200 µl
- Tip Style:Flat, O.D. 0.4 mm