ContactUsLinkComponent
Gel-Loading Pipet Tips
Corning® Gel-Loading Pipette Tips, Corning
# de catalogue 29442-666
Fournisseur:  Corning
Product available on GSA Advantage®
Gel-Loading Pipet Tips
Corning® Gel-Loading Pipette Tips, Corning
# de catalogue 29442-666
Fournisseur:  Corning
Product available on GSA Advantage®
Spécifications
  • Volume:
    0.1 - 200 µl
  • Tip Style:
    Round, O.D. 0.5 mm
  • Supplier No.:
    4853
Spécifications
A propos de cet article
These gel-loading pipette tips feature a capillary end that allows easy access into vertical and horizontal electrophoresis gels.
  • Disposable
  • Certified RNase-/DNase-free and non pyrogenic
Ordering information: Available in flat and round tip configurations. Flat tips are designed for use in 0.2 and 0.4 mm sequencing gels. Round tips are designed for use in SDS-PAGE 0.5 mm gels.
Customers Who Bought This Also Bought