ContactUsLinkComponent
Cap Insrt Cryogenic Wht Cs-500
Corning® Cap Inserts for Cryogenic Vials with Closures, Polypropylene, Corning
# de catalogue 29442-544
Fournisseur:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
Cap Insrt Cryogenic Wht Cs-500
Corning® Cap Inserts for Cryogenic Vials with Closures, Polypropylene, Corning
# de catalogue 29442-544
Fournisseur:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
Spécifications
  • Color:
    White
  • Description:
    Color-Coded Cap Inserts
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Cat. no.:
    29442-544
  • Supplier No.:
    2015
Spécifications
A propos de cet article
The white polypropylene cap insert provide color-coding for easy sample identification.
  • Inserts are packaed in resealable bags
  • Non-sterile
Customers Who Bought This Also Bought