Costar® Reagent Reservoirs, Polystyrene, Sterile, Corning
# de catalogue 29442-476
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Capacity:100 ml
- Sterility:Sterile
- Color:White
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Cat. no.:29442-476
- Supplier No.:4872
Spécifications
A propos de cet article
White PS, sterile, disposable.
- Ideal for repetitively filling multi channel pipettors
- Manufactured from virgin PS
Customers Who Bought This Also Bought