ContactUsLinkComponent
Costar* Reagent Reservoir
Costar® Reagent Reservoirs, Polystyrene, Sterile, Corning
# de catalogue 29442-476
Fournisseur:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
Costar® Reagent Reservoirs, Polystyrene, Sterile, Corning
# de catalogue 29442-476
Fournisseur:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
Spécifications
  • Capacity:
    100 ml
  • Sterility:
    Sterile
  • Color:
    White
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Cat. no.:
    29442-476
  • Supplier No.:
    4872
Spécifications
A propos de cet article
White PS, sterile, disposable.
  • Ideal for repetitively filling multi channel pipettors
  • Manufactured from virgin PS
Customers Who Bought This Also Bought