VWR® High Quality Bibulous Paper
Catalog # 28511-007
Supplier: VWR International
Specifications
- Description:VWR® high quality bibulous paper
- Size:50 sheets per block
- Cat. no.:28511-007
Specifications
About this item
High quality, highly absorbent paper manufactured for minimum linting and shedding.
- Neutral paper
- Lintless drying surface
Recommended for drying slides, and small parts or components. 50 sheets per book.
Customers Who Bought This Also Bought