ContactUsLinkComponent
VWR* High Quality Bibulous Paper
VWR® High Quality Bibulous Paper
Catalog # 28511-007
Supplier:  VWR International
VWR* High Quality Bibulous Paper
VWR® High Quality Bibulous Paper
Catalog # 28511-007
Supplier:  VWR International
Specifications
  • Description:
    VWR® high quality bibulous paper
  • Size:
    50 sheets per block
  • Cat. no.:
    28511-007
Specifications
About this item
High quality, highly absorbent paper manufactured for minimum linting and shedding.
  • Neutral paper
  • Lintless drying surface
Recommended for drying slides, and small parts or components. 50 sheets per book.
Customers Who Bought This Also Bought