Order Entry
ContactUsLinkComponent
BLACK FLTER PAPER
Black Qualitative Filter Paper, Grade 8613, Ahlstrom
# de catalogue 28342-052
Fournisseur:  Ahlstrom
BLACK FLTER PAPER
Black Qualitative Filter Paper, Grade 8613, Ahlstrom
# de catalogue 28342-052
Fournisseur:  Ahlstrom

Spécifications

  • Ø:
    4.7 cm
  • Flow rate:
    Medium
  • Retention:
    4 µm
  • Load capacity:
    Medium
  • Cat. no.:
    28342-052
  • Supplier No.:
    8613-0471

Spécifications

A propos de cet article

Ahlstrom 8613 has a smooth black finish designed for use in applications requiring visual examination of particles.

Customers Who Bought This Also Bought