VWR® Blotting Paper, Grade 703
46 x 57 cm (18 1/8 x 22 7/16"), 0.38 mm, Sheet
Supplier: VWR International
VWR® Blotting Paper, Grade 703
Catalog # 28298-020
Supplier: VWR International
Product available on GSA Advantage®
Jump to:
Product Details & Documents
Grade 703 blotting papers are ideal for use as wicks in protein and nucleic acid blotting.
- Made of 100% cotton fiber
- Uniform and smooth
- Produced using ultrapure water
- Contains no additives
Ideal for use as wicks in protein and nucleic acid blotting. Provides a uniform flow of buffer through the gel to the transfer membrane in a blotting sandwich. Also suitable for use in removing gels from glass supports.
Ordering information: Available cut to size, preventing the development of air pockets between the gel and transfer membrane. For more information, contact your Avantor sales representative.
Spécifications
- Type:Sheet
- Dimensions:46 x 57 cm (18 1/8 x 22 7/16")
- Épaisseur:0.38 mm
- Thickness:0.38 mm
- Cat. No.:28298-020
- Klemm wicking rate test:626 sec./7.5 cm rise
- Herzberg flow rate:250 sec./100 mL at 10 cm head of water
- Weight:185 g/m²
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



